Learn More
Cayman Chemical Dihydromyristicin, Purity: ≥95%, CAS: 52811-28-6, 10 mg
SDP
Catalog No. 502917175
Encompass_Preferred
A derivative of myristicin; induces GST activity in the liver and small intestinal mucosa but not the forestomach, lungs, large intestinal mucosa, or bladder in mice at 20 mg/animal
Catalog No. 50-291-7175 Supplier Cayman Chemical Supplier No. 3927210 MG
A derivative of myristicin; induces GST activity in the liver and small intestinal mucosa but not the forestomach, lungs, large intestinal mucosa, or bladder in mice at 20 mg/animal