Learn More
Cayman Chemical DIhydromyrIstIcIn 50mg
SDP
Catalog No. 502918473
Encompass_Preferred
A derivative of myristicin; induces GST activity in the liver and small intestinal mucosa but not the forestomach, lungs, large intestinal mucosa, or bladder in mice at 20 mg/animal
Catalog No. 50-291-8473 Supplier Cayman Chemical Supplier No. 3927250 MG
A derivative of myristicin; induces GST activity in the liver and small intestinal mucosa but not the forestomach, lungs, large intestinal mucosa, or bladder in mice at 20 mg/animal