Learn More
Cayman Chemical Dihydronarwedine, Purity: ≥90%, CAS: 21041-10-1, 10 mg
SDP
Catalog No. 502914006
Encompass_Preferred
An alkaloid and GSK3β inhibitor; inhibits recombinant human GSK3β at 10 µM
Catalog No. 50-291-4006 Supplier Cayman Chemical Supplier No. 3540910 MG
An alkaloid and GSK3β inhibitor; inhibits recombinant human GSK3β at 10 µM