Learn More
Cayman Chemical DIhydropyrenophorIn 100ug
SDP
Catalog No. 502946788
Encompass_Preferred
A macrolide fungal metabolite; derivative of pyrenophorin; herbicidal against S. halepense when used at a concentration of 0.065 µM; phytotoxic to a variety of agricultural crops, including barley, sorghum, wheat, and oats, as well as grasses, including goosegrass and Bermuda grass, in leaf puncture assays
Catalog No. 50-294-6788 Supplier Cayman Chemical Supplier No. 33536100 UG
A macrolide fungal metabolite; derivative of pyrenophorin; herbicidal against S. halepense when used at a concentration of 0.065 µM; phytotoxic to a variety of agricultural crops, including barley, sorghum, wheat, and oats, as well as grasses, including goosegrass and Bermuda grass, in leaf puncture assays