Learn More
Cayman Chemical Dihydroergocristine (mesylate), Purity: ≥95%, CAS: 24730-10-7, 100 mg
SDP
Catalog No. 502988141
Encompass_Preferred
An alkaloid with diverse biological activities; inhibits contraction of isolated rat vas deferens induced by norepinephrine (pA2 = 7.88); reduces blood pressure in a rat model of norepinephrine-induced hypertension and increases blood pressure in hypotensive rats from 0.1-100 µg/kg, i.v.; reduces aqueous humor production and intraocular pressure in a rabbit model of α-chymotrypsin-induced ocular hypertension; reduces hypermotility induced by ethanol in mice
Catalog No. 50-298-8141 Supplier Cayman Chemical Supplier No. 29492100 MG
An alkaloid with diverse biological activities; inhibits contraction of isolated rat vas deferens induced by norepinephrine (pA2 = 7.88); reduces blood pressure in a rat model of norepinephrine-induced hypertension and increases blood pressure in hypotensive rats from 0.1-100 µg/kg, i.v.; reduces aqueous humor production and intraocular pressure in a rabbit model of α-chymotrypsin-induced ocular hypertension; reduces hypermotility induced by ethanol in mice