Learn More
Cayman Chemical DIhydrorhodamIn 123 10mg
SDP
Catalog No. 502896307
Encompass_Preferred
A fluorophore that can be used as an indicator of peroxynitrite formation; neither NO, superoxide, nor hydrogen peroxide alone appear to oxidize DHR; used to investigate reactive oxygen intermediates produced by endothelial cells, eosinophils, and reactive microglia
Catalog No. 50-289-6307 Supplier Cayman Chemical Supplier No. 8510010 MG
A fluorophore that can be used as an indicator of peroxynitrite formation; neither NO, superoxide, nor hydrogen peroxide alone appear to oxidize DHR; used to investigate reactive oxygen intermediates produced by endothelial cells, eosinophils, and reactive microglia