Learn More
AdipoGen Dihydrorhodamine 123, 109244-58-8, MFCD04221428, 10 mg
SDP
Supplier:  AdipoGen CDXD0134M010
Encompass_Preferred
MF: C21H18N2O3, MW: 346.38, ≥95%, Appearance: Pink solid. Cell-permeable non-fluorescent reactive oxygen species (ROS) indicator. Passively diffuses across membranes where it is oxidized by peroxynitrite to rhodamine 123 which localizes in the mitochondria and exhibits green fluorescence (Ex/Em wavelengths of 500 and 536 nm). Used to investigate reactive oxygen intermediates produced by human and murine phagocytes, activated rat mast cells and cultured endothelial cells.
Catalog No. 50-595-606