Learn More
Enzo Life Sciences Dihydrorhodamine 123 (ultra pure) (10mg). CAS: 109244-58-8
SDP
Catalog No. 502012630
Encompass_Preferred
Dihydrorhodamine 123 is a widely utilized probe for detection of intracellular reactive oxygen species (ROS) such as peroxide, hypochlorous acid and peroxynitrite. It is readily oxidized into rhodamine 123, which exhibits a spectral profile similar to that of FITC. In combination with other fluorescent probes (such as surface receptor-targeted fluorescent antibodies, the cell viability probe propidium iodide, or fluorescent calcium indicators) dihydrorhodamine 123 can be used for multiparametric cell measurements. Wavelength Maxima: Excitation 507nm, Emission 529nm Alternative name: 2-(3,6-Diamino-9H-xanthene-9-yl)-benzoic acid methyl ester. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2630 Supplier Enzo Life Sciences Supplier No. ENZ52302
Dihydrorhodamine 123 is a widely utilized probe for detection of intracellular reactive oxygen species (ROS) such as peroxide, hypochlorous acid and peroxynitrite. It is readily oxidized into rhodamine 123, which exhibits a spectral profile similar to that of FITC. In combination with other fluorescent probes (such as surface receptor-targeted fluorescent antibodies, the cell viability probe propidium iodide, or fluorescent calcium indicators) dihydrorhodamine 123 can be used for multiparametric cell measurements. Wavelength Maxima: Excitation 507nm, Emission 529nm Alternative name: 2-(3,6-Diamino-9H-xanthene-9-yl)-benzoic acid methyl ester. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.