Learn More
Abcam Dihydrorhodamine 123, Fluorescent probe for ROS, 2MG
Supplier:  Abcam AB1452602MG
Encompass_Preferred
MW 346.4 Da, Purity >98%. Cell-permeable fluorescent probe for ROS. Redox sensor. Detects presence of peroxynitrite. Fluoresces brighter extracellularly than Dihydrofluorescein diacetate (ab145351) but less bight intracellularly.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-122