Learn More
TARGETMOL CHEMICALS INC DIHYDROSANGUINARINE 5MG
SDP
Catalog No. 502579828
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dihydrosanguinarine is a natural compound isolated from the leaves of Macleaya microcarpaand has antifungal and anticancer activity. purity: 99%
Catalog No. 50-257-9828 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S20125MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dihydrosanguinarine is a natural compound isolated from the leaves of Macleaya microcarpaand has antifungal and anticancer activity. purity: 99%