Learn More
Medchemexpress LLC Dihydrosinapyl alcohol | 20736-25-8 | MFCD00016572 | 98.1% | 212.24 g·mol⁻¹ | C11H16O4 | 1 ML
SDP
Catalog No. 5000301744
Encompass_Preferred
Dihydrosinapyl alcohol is a lignin-derived phenylpropanoid alcohol used as a research chemical and analytical reference. It is provided as a solid and as a 10 mM solution in DMSO and is employed in studies of lignocellulose conversion, hydrogenation/hydrogenolysis products, and small-molecule analytical assays.
  • Natural product phenylpropanoid relevant to lignocellulose and biomass research.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • High purity (approximately 98.1%) suitable for analytical and research use.
  • Molecular weight 212.24 g·mol⁻¹ and chemical formula C11H16O4 for accurate calculations.
  • Supplied in small quantities suitable for analytical standards and method development.
Catalog No. 50-003-01744 Supplier Medchemexpress LLC Supplier No. HYW02537110MM/1ML
Dihydrosinapyl alcohol is a lignin-derived phenylpropanoid alcohol used as a research chemical and analytical reference. It is provided as a solid and as a 10 mM solution in DMSO and is employed in studies of lignocellulose conversion, hydrogenation/hydrogenolysis products, and small-molecule analytical assays.
  • Natural product phenylpropanoid relevant to lignocellulose and biomass research.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • High purity (approximately 98.1%) suitable for analytical and research use.
  • Molecular weight 212.24 g·mol⁻¹ and chemical formula C11H16O4 for accurate calculations.
  • Supplied in small quantities suitable for analytical standards and method development.