Learn More
Cayman Chemical DIhydrosnguInarIn 10mg
SDP
Catalog No. 502989516
Encompass_Preferred
A metabolite of sanguinarine with diverse biological activities; inhibits proliferation of PANC-1, SW 1990, and HPDE6c7 cancer cells at 10, 20, and 30 µM; reduces the levels of phosphorylated ERK and C-RAF, halts the cell cycle at the G0/G1 and G2/M phases, and induces apoptosis in PANC-1 cells
Catalog No. 50-298-9516 Supplier Cayman Chemical Supplier No. 3113910 MG
A metabolite of sanguinarine with diverse biological activities; inhibits proliferation of PANC-1, SW 1990, and HPDE6c7 cancer cells at 10, 20, and 30 µM; reduces the levels of phosphorylated ERK and C-RAF, halts the cell cycle at the G0/G1 and G2/M phases, and induces apoptosis in PANC-1 cells