Learn More
Selleck Chemical LLC Dihydrostreptomycin sulfate S4523-100mg
SDP
Catalog No. 503111383
Encompass_Preferred
Dihydrostreptomycin sulfate is an aminoglycoside antibiotic used to treat bacterial diseases in cattle pigs and sheep Dihydrostreptomycin is a derivative of streptomycin
Catalog No. 50-311-1383 Supplier Selleck Chemical LLC Supplier No. S4523100MG
Dihydrostreptomycin sulfate is an aminoglycoside antibiotic used to treat bacterial diseases in cattle pigs and sheep Dihydrostreptomycin is a derivative of streptomycin