Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrostreptomycin Sulfate, Laboratory Grade, G-Biosciences
SCI_ED

Catalog No. S822011
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S822011 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S822011 Supplier G-Biosciences Supplier No. RC994
Add to Cart
Add to Cart

Dihydrostreptomycin is an aminoglycoside antibiotic targeting gram-positive bacteria and Mycobacterium species

It is a derivative of streptomycin and commonly used to study aminoglycoside uptake. Dihydrostreptomycin sulfate is freely soluble in aqueous solution.

Chemical Identifiers

Molecular Weight (g/mol) 730.71 g/mol

Specifications

Boiling Point 641.09°C
Quantity 25 g
Percent Purity >98%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.