Learn More
Cayman Chemical Dihydrostreptomycin (sulfate), Purity: ≥95%, CAS: 5490-27-7, 250 g
SDP
Catalog No. 502941752
Encompass_Preferred
An aminoglycoside antibiotic and a derivative of streptomycin; active against B. subtilis, K. pneumoniae, S. marcescens, S. aureus, P. aeruginosa, and M. tuberculosis (MICs = 0.07, 0.08, 0.09, 0.06, 0.8, and 5.7 µg/ml, respectively); interferes with bacterial protein synthesis by binding the 30S ribosomal subunit; enhances phagocytosis and intracellular killing of E. coli by isolated mouse peritoneal macrophages; improves survival in a mouse model of S. aureus infection
Catalog No. 50-294-1752 Supplier Cayman Chemical Supplier No. 27879250 G
An aminoglycoside antibiotic and a derivative of streptomycin; active against B. subtilis, K. pneumoniae, S. marcescens, S. aureus, P. aeruginosa, and M. tuberculosis (MICs = 0.07, 0.08, 0.09, 0.06, 0.8, and 5.7 µg/ml, respectively); interferes with bacterial protein synthesis by binding the 30S ribosomal subunit; enhances phagocytosis and intracellular killing of E. coli by isolated mouse peritoneal macrophages; improves survival in a mouse model of S. aureus infection