Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrotanshinone I, G-Biosciences
SCI_ED

Catalog No. S973537
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S973537 10 mg
S000034 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S973537 Supplier G-Biosciences Supplier No. RC996
Add to Cart
Add to Cart

Dihydrotanshinone I is a compound extracted from the plant Salvie miltorrhiza Bunge. It is used as a gram positive antibacterial and anti-cancer compound.

In both in vivo and in vitro research, dihydrotanshinone I generated reactive oxygen species which trigger apoptosis in bacteria and cancer cells.

Chemical Identifiers

Molecular Weight (g/mol) 278.30 g/mol

Specifications

Boiling Point 479.2°C
Quantity 10 mg
Percent Purity >98%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.