Learn More
Abcam Dihydrotestosterone ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2839791X96TESTS
Encompass_Preferred
Dihydrotestosterone ELISA Kit is a Competitive ELISA kit for the measurement of Dihydrotestosterone in Saliva, Urine, Plasma, Milk, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1998781