Learn More
Cusabio Technology LLC Rat Dihydrotestosterone,DHT ELISA Kit
Catalog No. NC2952880
Encompass_Preferred
This Rat DHT ELISA Kit was designed for the quantitative measurement of Rat DHT protein in serum, plasma, tissue homogenates. It is a Competitive ELISA kit, its detection range is 10 pg/mL-2000 pg/mL and the sensitivity is 5 pg/mL. One kit contains 96 wells.
Catalog No. NC2952880 Supplier Cusabio Technology LLC Supplier No. CSBE07879R
This Rat DHT ELISA Kit was designed for the quantitative measurement of Rat DHT protein in serum, plasma, tissue homogenates. It is a Competitive ELISA kit, its detection range is 10 pg/mL-2000 pg/mL and the sensitivity is 5 pg/mL. One kit contains 96 wells.