Learn More
Crystal Chem Inc Dihydrotestosterone (DHT) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80961
Encompass
The Dihydrotestosterone (DHT) ELISA is a highly sensitive assay that is used to quantify levels of human DHT in serum. Highlights: Kit uses sample size of only 50 µL. High sensitivity of 6 pg/mL. Wide dynamic range of 25 - 2500 pg/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7867