Learn More
Medchemexpress LLC Dihydrouracil (5,6-Dihydrouracil) | 504-07-4 | MFCD00006029 | 99.9% | 114.10 | 100 G
SDP
Catalog No. 5000453440
Encompass_Preferred
Dihydrouracil (5,6-Dihydrouracil), a metabolite of Uracil, can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient. Uracil is the endogenous substrate for DPD, and Dihydrouracil is the product of Uracil. Multiple studies have shown an association between high endogenous Uracil levels or a low Dihydrouracil/Uracil ratio and severe fluoropyrimidine-associated toxicity.
  • Metabolite of Uracil
  • Can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient
  • For research use only
Catalog No. 50-004-53440 Supplier Medchemexpress LLC Supplier No. HYW012926100G
Dihydrouracil (5,6-Dihydrouracil), a metabolite of Uracil, can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient. Uracil is the endogenous substrate for DPD, and Dihydrouracil is the product of Uracil. Multiple studies have shown an association between high endogenous Uracil levels or a low Dihydrouracil/Uracil ratio and severe fluoropyrimidine-associated toxicity.
  • Metabolite of Uracil
  • Can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient
  • For research use only