Learn More
Medchemexpress LLC Dihydrouracil (5,6-Dihydrouracil) | 504-07-4 | MFCD00006029 | 99.9% | 114.10 | 25 G
SDP
Catalog No. 5000453457
Encompass_Preferred
Dihydrouracil (5,6-Dihydrouracil) is a metabolite of Uracil that can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient individuals. Uracil is the endogenous substrate for DPD, and Dihydrouracil is the product of Uracil. Studies have shown an association between high endogenous Uracil levels or a low Dihydrouracil/Uracil ratio and severe fluoropyrimidine-associated toxicity.
  • Can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient.
  • Endogenous metabolite.
  • Appearance: solid, white to off-white.
Catalog No. 50-004-53457 Supplier Medchemexpress LLC Supplier No. 25GHYW012926
Dihydrouracil (5,6-Dihydrouracil) is a metabolite of Uracil that can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient individuals. Uracil is the endogenous substrate for DPD, and Dihydrouracil is the product of Uracil. Studies have shown an association between high endogenous Uracil levels or a low Dihydrouracil/Uracil ratio and severe fluoropyrimidine-associated toxicity.
  • Can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient.
  • Endogenous metabolite.
  • Appearance: solid, white to off-white.