Learn More
Medchemexpress LLC Dihydrouracil | 504-07-4 | 114.10 | 50 G
SDP
Catalog No. 5000366749
Encompass_Preferred
Dihydrouracil (Standard) is the analytical standard of Dihydrouracil. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments. It can also be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient individuals. This product is for research use only and not sold to patients.
  • Analytical standard for dihydrouracil
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
  • Can be used as a marker for identification of DPD-deficient individuals
  • Intended for research and analytical applications
Catalog No. 50-003-66749 Supplier Medchemexpress LLC Supplier No. HYW01292650G
Add to Cart
Add to Cart
Dihydrouracil (Standard) is the analytical standard of Dihydrouracil. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments. It can also be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient individuals. This product is for research use only and not sold to patients.
  • Analytical standard for dihydrouracil
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
  • Can be used as a marker for identification of DPD-deficient individuals
  • Intended for research and analytical applications