Learn More
Cayman Chemical DIhydroxy Melphaln 10mg
SDP
Catalog No. 502941754
Encompass_Preferred
An inactive degradation product of melphalan; formed by the hydrolysis of melphalan via the intermediate monohydroxy melphalan in aqueous solutions, including cell culture medium and human plasma
Catalog No. 50-294-1754 Supplier Cayman Chemical Supplier No. 2785710 MG
An inactive degradation product of melphalan; formed by the hydrolysis of melphalan via the intermediate monohydroxy melphalan in aqueous solutions, including cell culture medium and human plasma