Learn More
Cayman Chemical DIhydroxyaceton PhosphATm 50mg
SDP
Catalog No. 502947984
Encompass_Preferred
A glycerolipid precursor; erythrocyte levels are increased in patients with triosephosphate isomerase deficiency
Catalog No. 50-294-7984 Supplier Cayman Chemical Supplier No. 3464150 MG
A glycerolipid precursor; erythrocyte levels are increased in patients with triosephosphate isomerase deficiency