Learn More
Medchemexpress LLC Dihydroxyacetone phosphate | 57-04-5 | 99.9% | 170.06 | 25 MG
SDP
Catalog No. 5000439265
Encompass_Preferred
Dihydroxyacetone phosphate (DHPA), a derivative of Dihydroxyacetone (DHA), is a crucial intermediate involved in key pathways like glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be utilized as a substrate and metabolic marker in biochemical research.
  • Participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle.
  • Can be used as a substrate and metabolic marker in biochemical research.
  • Can be utilized in fructose metabolism, triose, and glycation research.
  • Has potential glycating effects to form advanced glycation end products (AGEs).
Catalog No. 50-004-39265 Supplier Medchemexpress LLC Supplier No. 25MGHY113131
Dihydroxyacetone phosphate (DHPA), a derivative of Dihydroxyacetone (DHA), is a crucial intermediate involved in key pathways like glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be utilized as a substrate and metabolic marker in biochemical research.
  • Participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle.
  • Can be used as a substrate and metabolic marker in biochemical research.
  • Can be utilized in fructose metabolism, triose, and glycation research.
  • Has potential glycating effects to form advanced glycation end products (AGEs).