Learn More
Medchemexpress LLC Dihydroxyacetone phosphate | 57-04-5 | 99.9% | 170.06 | 50 MG
SDP
Catalog No. 5000437195
Encompass_Preferred
Dihydroxyacetone phosphate (DHAP) is a derivative of Dihydroxyacetone (DHA) and is an important intermediate that participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be used as a substrate and metabolic marker in biochemical research, and also has potential glycating effects to form advanced glycation end products (AGEs).
  • Can be used in fructose metabolism, triose and glycation research
  • Suitable as a substrate and metabolic marker in biochemical research
Catalog No. 50-004-37195 Supplier Medchemexpress LLC Supplier No. HY11313150MG
Dihydroxyacetone phosphate (DHAP) is a derivative of Dihydroxyacetone (DHA) and is an important intermediate that participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be used as a substrate and metabolic marker in biochemical research, and also has potential glycating effects to form advanced glycation end products (AGEs).
  • Can be used in fructose metabolism, triose and glycation research
  • Suitable as a substrate and metabolic marker in biochemical research