Learn More
Medchemexpress LLC Dihydroxyacetone phosphate | 57-04-5 | MFCD00871491 | 99.9% | 170.06 g/mol | C3H7O6P | 5 MG
SDP
Catalog No. 5000264334
Encompass_Preferred
Dihydroxyacetone phosphate (DHAP) is an endogenous metabolite and a key intermediate in glycolysis and lipid biosynthesis. It is commonly used as a research reagent in biochemical, enzymology, and metabolic pathway studies, including investigations of metabolic disorders.
  • High purity (99.9%) suitable for analytical and biochemical assays.
  • Molecular weight 170.06 g/mol; chemical formula C3H7O6P.
  • Available in small pack sizes for research use: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Appropriate for enzymology, metabolic pathway, and assay development studies.
  • Stable when stored per supplier recommendations; follow SDS for handling.
Catalog No. 50-002-64334 Supplier Medchemexpress LLC Supplier No. HY1131315MG
Dihydroxyacetone phosphate (DHAP) is an endogenous metabolite and a key intermediate in glycolysis and lipid biosynthesis. It is commonly used as a research reagent in biochemical, enzymology, and metabolic pathway studies, including investigations of metabolic disorders.
  • High purity (99.9%) suitable for analytical and biochemical assays.
  • Molecular weight 170.06 g/mol; chemical formula C3H7O6P.
  • Available in small pack sizes for research use: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Appropriate for enzymology, metabolic pathway, and assay development studies.
  • Stable when stored per supplier recommendations; follow SDS for handling.