Learn More
Medchemexpress LLC Dihydroxyacetone phosphate (DHAP) | 10 MG
SDP
Catalog No. 5000435607
Encompass_Preferred
Dihydroxyacetone phosphate (DHAP), a derivative of Dihydroxyacetone (DHA), is an important intermediate that participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be used as a substrate and metabolic marker in biochemical research.
  • Important intermediate in key pathways such as glycolysis, lipid biosynthesis, and the plant Calvin cycle.
  • Can be used as a substrate and metabolic marker in biochemical research.
Catalog No. 50-004-35607 Supplier Medchemexpress LLC Supplier No. HY11313110MG
Dihydroxyacetone phosphate (DHAP), a derivative of Dihydroxyacetone (DHA), is an important intermediate that participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be used as a substrate and metabolic marker in biochemical research.
  • Important intermediate in key pathways such as glycolysis, lipid biosynthesis, and the plant Calvin cycle.
  • Can be used as a substrate and metabolic marker in biochemical research.