Learn More
Medchemexpress LLC Dihydroxyacetone phosphate hemimagnesium hydrate | 00-00-0 | MFCD00871491 | 170.06 g·mol⁻¹ | C3H7O6P | 5 MG
SDP
Catalog No. 5000264048
Encompass_Preferred
Dihydroxyacetone phosphate hemimagnesium hydrate is a biochemical intermediate involved in glycolysis and lipid biosynthesis. Supplied as a small-scale research reagent, it is intended for laboratory use in metabolic pathway studies, enzyme assays, and synthetic biochemistry applications.
  • Used in studies of glycolysis and lipid biosynthesis.
  • Suitable for enzyme assays and metabolic pathway research.
  • Provided as a hemimagnesium hydrate salt to support solubility and handling.
  • Chemical formula C3H7O6P and molecular weight 170.06 g·mol⁻¹.
  • Available in small-scale quantities for experimental workflows.
Catalog No. 50-002-64048 Supplier Medchemexpress LLC Supplier No. HY113131A5MG
Dihydroxyacetone phosphate hemimagnesium hydrate is a biochemical intermediate involved in glycolysis and lipid biosynthesis. Supplied as a small-scale research reagent, it is intended for laboratory use in metabolic pathway studies, enzyme assays, and synthetic biochemistry applications.
  • Used in studies of glycolysis and lipid biosynthesis.
  • Suitable for enzyme assays and metabolic pathway research.
  • Provided as a hemimagnesium hydrate salt to support solubility and handling.
  • Chemical formula C3H7O6P and molecular weight 170.06 g·mol⁻¹.
  • Available in small-scale quantities for experimental workflows.