Learn More
Medchemexpress LLC Dihydroxyacetone phosphate hemimagnesium hydrate | 99.9% | 380.42 g/mol | 10 MG
SDP
Catalog No. 5000436143
Encompass_Preferred
Dihydroxyacetone phosphate (DHPA) hemimagnesium hydrate, a derivative of Dihydroxyacetone (DHA), is an important intermediate that participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be used as a substrate and metabolic marker in biochemical research.
  • Substrate and metabolic marker in biochemical research
  • Participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle
  • Can be used in fructose metabolism, triose, and glycation research
  • Potential glycating effects to form advanced glycation end products (AGEs)
Catalog No. 50-004-36143 Supplier Medchemexpress LLC Supplier No. HY113131A10MG
Dihydroxyacetone phosphate (DHPA) hemimagnesium hydrate, a derivative of Dihydroxyacetone (DHA), is an important intermediate that participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be used as a substrate and metabolic marker in biochemical research.
  • Substrate and metabolic marker in biochemical research
  • Participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle
  • Can be used in fructose metabolism, triose, and glycation research
  • Potential glycating effects to form advanced glycation end products (AGEs)