Learn More
American Radiolabeled Chemicals Inc DIHYDROXYFLAVONE 10ME
SDP
Catalog No. 50353078
Encompass
7,8-Dihydroxyflavone 10mg as solid, M.W. 254.24, Solvent: Solid, CAS # 38183-03-8, Tyrosine kinase receptor B (TrkB) agonist
Catalog No. 50-353-078 Supplier American Radiolabeled Chemicals Inc Supplier No. ARCD 074010MG
7,8-Dihydroxyflavone 10mg as solid, M.W. 254.24, Solvent: Solid, CAS # 38183-03-8, Tyrosine kinase receptor B (TrkB) agonist