Learn More
Enzo Life Sciences Dihydrosphingosine-1-phosphate, D-erythro (1 mg). CAS: 28508-07-8
SDP
Supplier:  Enzo Life Sciences BMLSL1430001
Encompass_Preferred
Saturated analog of sphingosine-1-phosphate. Induces chemotaxis in HUVECs. May be used as a negative control for intracellular effects of S1P. Rapidly dephosphorylated by a sphingosine phosphatase in cultured human fibroblasts. Ligand for S1P (EDG) receptors. Purity: ≥98%. Solubility: Soluble in methanol/THF/water (60/30/10) (0.5mg/ml). Slightly soluble in DMSO, methanol or acetic acid. Long Term Storage: -20°C.
Catalog No. 50-201-2119