Learn More
Medchemexpress LLC HY-D0083 10mg Medchemexpress, DiI CAS:41085-99-8 Purity:>98%
SDP
Catalog No. 501933146
Encompass_Preferred
Medchemexpress, HY-D0083 10mg DiI CAS:41085-99-8 DiI (DiIC18(3)) is a lipophilic membrane dye that diffuses laterally to stain the entire cell. DiI is excited to emit orange-red fluorescence. The fluorescence is weak in water but strong and photostable when incorporated into membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3146 Supplier Medchemexpress LLC Supplier No. HYD008310MG
Medchemexpress, HY-D0083 10mg DiI CAS:41085-99-8 DiI (DiIC18(3)) is a lipophilic membrane dye that diffuses laterally to stain the entire cell. DiI is excited to emit orange-red fluorescence. The fluorescence is weak in water but strong and photostable when incorporated into membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.