Learn More
AdipoGen DiI
SDP
Catalog No. 502561173
Encompass_Preferred
Chemical. CAS 41085-99-8. Formula C59H97ClN2O4. MW 933.87. Synthetic. Endoplasmic reticulum membrane stain. Lipophilic carbocyanine dye used primarily for optical recordings of membrane voltage and for studies of membrane fluidity. Retrograde stain for neurons provides intense, long-lasting staining of live neurons in vivo and in vitro.
Catalog No. 50-256-1173 Supplier AdipoGen Supplier No. CDXD0230G001
Chemical. CAS 41085-99-8. Formula C59H97ClN2O4. MW 933.87. Synthetic. Endoplasmic reticulum membrane stain. Lipophilic carbocyanine dye used primarily for optical recordings of membrane voltage and for studies of membrane fluidity. Retrograde stain for neurons provides intense, long-lasting staining of live neurons in vivo and in vitro.