Learn More
Cell Applications Inc DII-AC-LDL KIT
SDP
Catalog No. 50608745
Encompass
DiI-conjugated acetylated low-density lipoproteins can be used to identify endothelial cells.
Catalog No. 50-608-745 Supplier Cell Applications Inc Supplier No. 022K
DiI-conjugated acetylated low-density lipoproteins can be used to identify endothelial cells.