Learn More
Abcam DiI, Endoplasmic reticulum membrane stain, 100MG
Catalog No. 502705298
Encompass_Preferred
MW 933.87 g/mol, Purity >98%. Lipophilic carbocyanine dye used primarily for optical recordings of membrane voltage and for studies of membrane fluidity. Retrograde stain for neurons; provides intense, long-lasting staining of live neurons in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5298 Supplier Abcam Supplier No. AB145311100MG
MW 933.87 g/mol, Purity >98%. Lipophilic carbocyanine dye used primarily for optical recordings of membrane voltage and for studies of membrane fluidity. Retrograde stain for neurons; provides intense, long-lasting staining of live neurons in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement