Learn More
Kalen Biomedical LLC DII-HDL .5MG
Supplier:  Kalen Biomedical LLC 7703309
Encompass_Preferred
Purified human HDL is labeled with DiI, a lipophylic fluorescent dye which can be excited at 488nm or 514nm with an emission at 565 nm. The product is provided in PBS with EDTA as a preservative. The protein concentration is 0.5 mg/mL.
Catalog No. NC0341664