Learn More
Aat Bioquest DiI iodide [11-Dioctadecyl-3333- tetramethylindocarbocyanine iodide] (100 mg)
SDP
Supplier:  Aat Bioquest 22101
Encompass_Preferred
DiI DiO DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures. This catalog is for 100 mg.
Catalog No. 50-225-0050