Learn More
Kalen Biomedical LLC DII LABELED NATIVE LDL
Catalog No. NC9839048
Encompass_Preferred
Purified human LDL is labeled with DiI, a lipophylic fluorescent dye which can be excited at 488nm or 514nm with an emission at 565 nm. The product is provided in PBS with EDTA as a preservative. The protein concentration is 0.5 mg/mL.
Catalog No. NC9839048 Supplier Kalen Biomedical LLC Supplier No. 7702309
Purified human LDL is labeled with DiI, a lipophylic fluorescent dye which can be excited at 488nm or 514nm with an emission at 565 nm. The product is provided in PBS with EDTA as a preservative. The protein concentration is 0.5 mg/mL.