Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DiI-Lipoprotein, High Density, Human Plasma

Catalog No. AAJ65503MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65503MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65503MCR Supplier Thermo Scientific™ Supplier No. J65503MCR

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

For research use

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
Formulation Liquid
Quantity 1 mg
Source Human plasma
Storage Requirements 2° to 8°C
Regulatory Status RUO
Protein Length 3,3'-Dioctadecylindocarbocyanine-high density lipoprotein
Product Type DiI-Lipoprotein, high density, Human Plasma
Recombinant Native
Form Liquid

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.