Learn More
Aat Bioquest DiI perchlorate [11-Dioctadecyl-3333-tetramethylindocarbocyanine perchlorate] CAS 41085-99-8 (100 mg)
SDP
Catalog No. 502250051
Encompass_Preferred
DiI DiO DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures. This catalog is for 100 mg.
This item is not returnable. View return policy
Catalog No. 50-225-0051 Supplier Aat Bioquest Supplier No. 22102
Add to Cart
Edge
Add to Cart
DiI DiO DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures. This catalog is for 100 mg.