Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diiodo(p-cymene)ruthenium(II) dimer | 90614-07-6 | 98.0% | C20H28I4Ru2 | 1 G
SDP

Catalog No. 5000433286
Encompass_Preferred

Diiodo(p-cymene)ruthenium(II) dimer is a chemical compound used in laboratory settings and for the manufacture of various substances. This product presents as a brown to reddish brown solid, known for its chemical stability under recommended storage conditions.

  • Appearance: Brown to reddish brown solid
  • Molecular weight: 978.19
  • Purity (NMR): 98.0%
  • Recommended storage conditions: 4°C, protect from light, stored under nitrogen
  • Melting/freezing point: 234-236 °C

Catalog No. 50-004-33286 Supplier Medchemexpress LLC Supplier No. HY799861G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diiodo(p-cymene)ruthenium(II) dimer is a chemical compound used in laboratory settings and for the manufacture of various substances. This product presents as a brown to reddish brown solid, known for its chemical stability under recommended storage conditions.

  • Appearance: Brown to reddish brown solid
  • Molecular weight: 978.19
  • Purity (NMR): 98.0%
  • Recommended storage conditions: 4°C, protect from light, stored under nitrogen
  • Melting/freezing point: 234-236 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.