Learn More
Medchemexpress LLC Diiodo(p-cymene)ruthenium(II) dimer | 90614-07-6 | 98.0% | C20H28I4Ru2 | 25 G
SDP
Catalog No. 5000358574
Encompass_Preferred
Diiodo(p-cymene)ruthenium(II) dimer is a versatile ruthenium complex that serves as a drug intermediate for the synthesis of various active compounds. This air-stable, dimeric ruthenium complex is a precursor for a wide array of monomeric and dimeric ruthenium(II) complexes and acts as a catalyst for numerous reactions, including ruthenium-catalyzed hydrogenation.
  • Drug intermediate for active compound synthesis
  • Air-stable dimeric ruthenium complex
  • Precursor for various ruthenium(II) complexes
  • Catalyst for ruthenium-catalyzed hydrogenation
Catalog No. 50-003-58574 Supplier Medchemexpress LLC Supplier No. 25GHY79986
Diiodo(p-cymene)ruthenium(II) dimer is a versatile ruthenium complex that serves as a drug intermediate for the synthesis of various active compounds. This air-stable, dimeric ruthenium complex is a precursor for a wide array of monomeric and dimeric ruthenium(II) complexes and acts as a catalyst for numerous reactions, including ruthenium-catalyzed hydrogenation.
  • Drug intermediate for active compound synthesis
  • Air-stable dimeric ruthenium complex
  • Precursor for various ruthenium(II) complexes
  • Catalyst for ruthenium-catalyzed hydrogenation