Learn More
Medchemexpress LLC Diiodohydroxyquinoline | 83-73-8 | 98.0% | 1 ML
SDP
Catalog No. 5000412782
Encompass_Preferred
Diiodohydroxyquinoline (Iodoquinol) is a compound with orally active antiparasitic properties. It exhibits potent anti-SARS-CoV-2 activity with an EC50 value of 1.38 μM in VeroE6 cells, and its antimutagen is ascorbic acid. This compound shows promise for research in inflammation, intestinal amebiasis, amebic liver abscess, and chronic nonspecific diarrheas.
  • Possesses orally active antiparasitic properties.
  • Exhibits mutagenic potential in mice.
  • Shows potent anti-SARS-CoV-2 activity with an EC50 of 1.38 μM in VeroE6 cells.
  • Antimutagen is ascorbic acid.
  • Useful for research in inflammation.
  • Useful for research in intestinal amebiasis.
  • Useful for research in amebic liver abscess.
  • Useful for research in chronic nonspecific diarrheas.
Catalog No. 50-004-12782 Supplier Medchemexpress LLC Supplier No. HYB140010MM/1ML
Diiodohydroxyquinoline (Iodoquinol) is a compound with orally active antiparasitic properties. It exhibits potent anti-SARS-CoV-2 activity with an EC50 value of 1.38 μM in VeroE6 cells, and its antimutagen is ascorbic acid. This compound shows promise for research in inflammation, intestinal amebiasis, amebic liver abscess, and chronic nonspecific diarrheas.
  • Possesses orally active antiparasitic properties.
  • Exhibits mutagenic potential in mice.
  • Shows potent anti-SARS-CoV-2 activity with an EC50 of 1.38 μM in VeroE6 cells.
  • Antimutagen is ascorbic acid.
  • Useful for research in inflammation.
  • Useful for research in intestinal amebiasis.
  • Useful for research in amebic liver abscess.
  • Useful for research in chronic nonspecific diarrheas.