Learn More
Medchemexpress LLC Diiodohydroxyquinoline | 83-73-8 | 98.0% | 500 MG
SDP
Catalog No. 5000412566
Encompass_Preferred
Diiodohydroxyquinoline (also known as Iodoquinol) is an orally active compound with antiparasitic properties. It has shown potent anti-SARS-CoV-2 activity and mutagenic potential in mice, with ascorbic acid acting as an antimutagen. This compound is promising for research in inflammation, intestinal amebiasis, amebic liver abscess, and chronic nonspecific diarrheas.
  • Orally active with antiparasitic properties
  • Exhibits mutagenic potential in mice
  • Potent anti-SARS-CoV-2 activity with an EC50 value of 1.38 μM in VeroE6 cells
  • Antimutagenic effect can be mitigated by ascorbic acid
  • Suitable for research in inflammation
  • Suitable for research in intestinal amebiasis
  • Suitable for research in amebic liver abscess
  • Suitable for research in chronic nonspecific diarrheas
Catalog No. 50-004-12566 Supplier Medchemexpress LLC Supplier No. HYB1400500MG
Diiodohydroxyquinoline (also known as Iodoquinol) is an orally active compound with antiparasitic properties. It has shown potent anti-SARS-CoV-2 activity and mutagenic potential in mice, with ascorbic acid acting as an antimutagen. This compound is promising for research in inflammation, intestinal amebiasis, amebic liver abscess, and chronic nonspecific diarrheas.
  • Orally active with antiparasitic properties
  • Exhibits mutagenic potential in mice
  • Potent anti-SARS-CoV-2 activity with an EC50 value of 1.38 μM in VeroE6 cells
  • Antimutagenic effect can be mitigated by ascorbic acid
  • Suitable for research in inflammation
  • Suitable for research in intestinal amebiasis
  • Suitable for research in amebic liver abscess
  • Suitable for research in chronic nonspecific diarrheas