Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diiodomethane (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. D0610100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0610100G 100 g
D061025G 25 g
D0610500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0610100G Supplier TCI America Supplier No. D0610100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-11-6
Molecular Formula CH2I2
Molecular Weight (g/mol) 267.84
MDL Number MFCD00001079
InChI Key NZZFYRREKKOMAT-UHFFFAOYSA-N
Synonym methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
PubChem CID 6346
IUPAC Name diiodomethane
SMILES ICI

Specifications

Color Yellow
Boiling Point 181°C
Quantity 100 g
UN Number 1760
Formula Weight 267.84
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diiodomethane (stabilized with Copper chip)
EINECSNumber (2)-0043
RTECSNumber PA8575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.