Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisoamylamine 97.0+%, TCI America™
SDP

Catalog No. D061325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D061325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D061325ML Supplier TCI America Supplier No. D061325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 544-00-3
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.301
MDL Number MFCD00015053
InChI Key SPVVMXMTSODFPU-UHFFFAOYSA-N
Synonym diisopentylamine, diisoamylamine, 1-butanamine, 3-methyl-n-3-methylbutyl, di 3-methylbutyl amine, bis 3-methylbutyl amine, di-iso-pentylamine = di-iso-amylamine, 3-methyl-n-3-methylbutyl butan-1-amine, diisoamylamin, di-iso-amylamine, acmc-1astz
PubChem CID 10988
IUPAC Name 3-methyl-N-(3-methylbutyl)butan-1-amine
SMILES CC(C)CCNCCC(C)C

Specifications

Color Yellow
Boiling Point 187°C
Quantity 25 mL
UN Number 2924
Formula Weight 157.30
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diisoamylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.