Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diisobutyl adipate

Catalog No. 450588250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-058-8250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-058-8250ML Supplier Sigma Aldrich Supplier No. 450588250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diisobutyl adipate is a bis(2-methylpropyl) ester of adipic acid that is used as a plasticizer and a fatty acid in synthetic oils.

Specifications

CAS 141-04-8
Density 0.954 g/mL (at 25°C)
Boiling Point 293°C
Molecular Formula C14H26O4
Refractive Index n20/D 1.432 (literature)
Linear Formula [-CH2CH2CO2CH2CH(CH3)2]2
Quantity 250 mL
MDL Number MFCD00053722
Synonym Hexanedioic acid 1,6-bis(2-methylpropyl) ester; NSC 6343
Molecular Weight (g/mol) 258.35
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.