Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diisobutyl hydrogen phosphate | 6303-30-6 | MFCD25965560 | 98.0% | 210.21 | C8H19O4P | 10 MG
SDP

Catalog No. 5000284226
Encompass_Preferred

Diisobutyl hydrogen phosphate is an organophosphate compound used primarily as a flame retardant and as a reagent in biochemical assays. It has molecular formula C8H19O4P, molecular weight 210.21 g/mol, and CAS number 6303-30-6. Supplied in small research quantities, users should refer to the certificate of analysis for batch-specific handling and purity information.

  • Used as a flame retardant and biochemical reagent.
  • Molecular formula C8H19O4P and molecular weight 210.21 g/mol.
  • Provided in small research quantities suitable for assay development.
  • Purity guidance value 98.0%; actual purity may vary by batch.
  • Store under conditions specified in the certificate of analysis.

Catalog No. 50-002-84226 Supplier Medchemexpress LLC Supplier No. HYW58588410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diisobutyl hydrogen phosphate is an organophosphate compound used primarily as a flame retardant and as a reagent in biochemical assays. It has molecular formula C8H19O4P, molecular weight 210.21 g/mol, and CAS number 6303-30-6. Supplied in small research quantities, users should refer to the certificate of analysis for batch-specific handling and purity information.

  • Used as a flame retardant and biochemical reagent.
  • Molecular formula C8H19O4P and molecular weight 210.21 g/mol.
  • Provided in small research quantities suitable for assay development.
  • Purity guidance value 98.0%; actual purity may vary by batch.
  • Store under conditions specified in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.