Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diisobutyl hydrogen phosphate | 6303-30-6 | MFCD25965560 | 98.0% | 210.21 | C8H19O4P | 25 MG
SDP

Catalog No. 5000284227
Encompass_Preferred

Diisobutyl hydrogen phosphate is an organophosphate compound described as a flame retardant and supplied as a small-quantity laboratory reagent. It is reported with molecular formula C8H19O4P, molecular weight 210.21, and CAS number 6303-30-6. Typical pack sizes include 5 mg, 10 mg, and 25 mg; vendor listings report an approximate purity of 98%.

  • Organophosphate flame retardant used in material testing and research.
  • Available in small, research-friendly pack sizes suitable for analytical work.
  • Molecular formula C8H19O4P and molecular weight 210.21 facilitate formulation and calculations.
  • CAS number 6303-30-6 enables unambiguous substance identification.
  • Reported purity around 98%; verify with certificate of analysis for critical applications.

Catalog No. 50-002-84227 Supplier Medchemexpress LLC Supplier No. HYW58588425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diisobutyl hydrogen phosphate is an organophosphate compound described as a flame retardant and supplied as a small-quantity laboratory reagent. It is reported with molecular formula C8H19O4P, molecular weight 210.21, and CAS number 6303-30-6. Typical pack sizes include 5 mg, 10 mg, and 25 mg; vendor listings report an approximate purity of 98%.

  • Organophosphate flame retardant used in material testing and research.
  • Available in small, research-friendly pack sizes suitable for analytical work.
  • Molecular formula C8H19O4P and molecular weight 210.21 facilitate formulation and calculations.
  • CAS number 6303-30-6 enables unambiguous substance identification.
  • Reported purity around 98%; verify with certificate of analysis for critical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.